missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylquinoxaline, 97%, Thermo Scientific Chemicals

Catalog No. AAH2668003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2668003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2668003 Supplier Thermo Scientific Chemicals Supplier No. H2668003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 25594-62-1 | C10H8N2O | 172.187 g/mol

2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylquinoxaline is used as dyes, pharmaceuticals, and antibiotics such as olaquindox, carbadox, echinomycin, levomycin and actinoleutin. It is also used to make amiloride.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 25594-62-1
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
MDL Number MFCD07808559
InChI Key SWGILLQQUULMJB-UHFFFAOYSA-N
Synonym 2-acetylquinoxaline, 1-quinoxalin-2-yl ethanone, ethanone, 1-2-quinoxalinyl, acmc-1cknl, 1-quinoxalin-2-yl ethan-1-one
PubChem CID 11105814
IUPAC Name 1-quinoxalin-2-ylethanone
SMILES CC(=O)C1=NC2=CC=CC=C2N=C1

Specifications

Melting Point 76°C to 79°C
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 2-Acetylquinoxaline
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only