missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrrole, 99%

Catalog No. AC252490050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC252490050 5 g Glass bottle
AC252490250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC252490050 Supplier Thermo Scientific Chemicals Supplier No. 252490050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1072-83-9 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1072-83-9
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00005220
InChI Key IGJQUJNPMOYEJY-UHFFFAOYSA-N
Synonym 2-acetylpyrrole, 1-1h-pyrrol-2-yl ethanone, 1-1h-pyrrol-2-yl ethan-1-one, 2-acetyl pyrrole, 2-pyrrolylethanone, 2-acetyl-1h-pyrrole, methyl pyrrol-2-yl ketone, ethanone, 1-1h-pyrrol-2-yl, 2-pyrrolyl methyl ketone, ketone, methyl pyrrol-2-yl
PubChem CID 14079
ChEBI CHEBI:59981
IUPAC Name 1-(1H-pyrrol-2-yl)ethanone
SMILES CC(=O)C1=CC=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White to Beige
Boiling Point 220°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 271
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents,soluble in ethanol and ether (20°C),soluble in alcohol and propylene glycol
Formula Weight 109.13
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylpyrrole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 214-016-2

RUO – Research Use Only