missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyridine, 98%

Catalog No. p-7020655
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1268814 25 g
AAA1268822 100 g
AAA1268836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1268814 Supplier Thermo Scientific Chemicals Supplier No. A1268814
Only null left

CAS: 1122-62-9 | C7H7NO | 121.139 g/mol

2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylpyridine used as flavoring agent. It is also used in application for food additive.

Solubility
Soluble in water (18.2 g/100g @ 25C). Soluble in and acetate. Slightly soluble in carbon tetrachloride.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.139
MDL Number MFCD00006303
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1

Specifications

Melting Point 8°C to 10°C
Density 1.082
Boiling Point 189°C to 190°C
Flash Point 76°C (169°F)
Odor Unpleasant
Refractive Index 1.522
Quantity 25 g
Beilstein 107759
Solubility Information Soluble in water (18.2g/100g @ 25C). Soluble in and acetate. Slightly soluble in carbon tetrachloride.
Formula Weight 121.14
Percent Purity 98%
Chemical Name or Material 2-Acetylpyridine
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 214-355-6
RTECSNumber OB5310000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only