missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylpyrazine 99.0+%, TCI America™
$154.71 - $303.84
Chemical Identifiers
| CAS | 22047-25-2 |
|---|---|
| Molecular Formula | C6H6N2O |
| Molecular Weight (g/mol) | 122.127 |
| MDL Number | MFCD00006134 |
| InChI Key | DBZAKQWXICEWNW-UHFFFAOYSA-N |
| Synonym | acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone |
| PubChem CID | 30914 |
| IUPAC Name | 1-pyrazin-2-ylethanone |
| SMILES | CC(=O)C1=NC=CN=C1 |
Chemical Identifiers
| 22047-25-2 | |
| 122.127 | |
| DBZAKQWXICEWNW-UHFFFAOYSA-N | |
| 30914 | |
| CC(=O)C1=NC=CN=C1 |
| C6H6N2O | |
| MFCD00006134 | |
| acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone | |
| 1-pyrazin-2-ylethanone |
Specifications
| 22047-25-2 | |
| 72°C | |
| MFCD00006134 | |
| acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone | |
| CC(=O)C1=NC=CN=C1 | |
| 122.127 | |
| 122.13 | |
| Crystalline Powder |
| 77°C | |
| C6H6N2O | |
| 10 g | |
| DBZAKQWXICEWNW-UHFFFAOYSA-N | |
| 1-pyrazin-2-ylethanone | |
| 30914 | |
| ≥99.0% (GC) | |
| 2-Acetylpyrazine |
Safety and Handling
TSCA : Yes