missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetylpyrazine 99.0+%, TCI America™
Specifications
Specifications
| Melting Point | 77°C |
| Boiling Point | 72°C |
| Quantity | 10 g |
| Formula Weight | 122.13 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Acetylpyrazine |
Safety and Handling
| TSCA | Yes |