missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylpyrazine, 99%

CAS: 22047-25-2 | C6H6N2O | 122.127 g/mol

Supplier:  Thermo Scientific Chemicals A1591706

 View more versions of this product

Catalog No. AAA1591706


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Acetylpyrazine
75°C to 78°C
C6H6N2O
5 g
acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone
CC(=O)C1=NC=CN=C1
122.127
122.13
22047-25-2
78°C to 79°C (8 mmHg)
MFCD00006134
109630
DBZAKQWXICEWNW-UHFFFAOYSA-N
1-pyrazin-2-ylethanone
30914
99%

Chemical Identifiers

22047-25-2
122.127
DBZAKQWXICEWNW-UHFFFAOYSA-N
30914
CC(=O)C1=NC=CN=C1
C6H6N2O
MFCD00006134
acetylpyrazine, 2-acetylpyrazine, ethanone, 1-pyrazinyl, 1-pyrazin-2-yl ethan-1-one, methyl pyrazinyl ketone, 2-acetyl pyrazine, ketone, methyl pyrazinyl, 1-pyrazinylethanone, 1-pyrazin-2-yl-ethanone, 1-pyrazin-2-yl ethanone
1-pyrazin-2-ylethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 244-753-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only