missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylcyclopentanone 95.0+%, TCI America™

Catalog No. A086925ML
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
5 mL
Catalog No. Quantity
A086925ML 25 mL
A08695ML 5 mL
2 options

Catalog No. A086925ML

Supplier: TCI America A086925ML

Only null left

Chemical Identifiers

CAS 1670-46-8
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001413
InChI Key OSWDNIFICGLKEE-UHFFFAOYNA-N
Synonym 2-acetylcyclopentanone, cyclopentanone, 2-acetyl, o-acetylcyclopentanone, alpha-acetylcyclopentanone, acetylcyclopentanone, 2-acetyl cyclopentanone, 2-acetyl-cyclopentanon, 2-acetyl-cyclopentanone, acmc-209dvh, cyclopentanone,2-acetyl
PubChem CID 98471
IUPAC Name 2-acetylcyclopentan-1-one
SMILES CC(=O)C1CCCC1=O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 mL
Formula Weight 126.16
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylcyclopentanone
RTECSNumber GY4728000
TSCA Yes