missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylcyclohexanone 97.0+%, TCI America™

Catalog No. p-7117106
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A088110ML 10 mL
A088125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A088110ML Supplier TCI America Supplier No. A088110ML
Only null left

Chemical Identifiers

CAS 874-23-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001633
InChI Key OEKATORRSPXJHE-UHFFFAOYNA-N
Synonym 2-acetylcyclohexanone, cyclohexanone, 2-acetyl, 2-acetyl-1-cyclohexanone, .alpha.-acetylcyclohexanone, 2-acetyl cyclohexanone, 2-acetyl-cyclohexanone, cyclohexanone,2-acetyl, 1-acetyl-2-oxocyclohexane, alpha-acetylcyclohexanone, acetylcyclohexanone
PubChem CID 13400
IUPAC Name 2-acetylcyclohexan-1-one
SMILES CC(=O)C1CCCCC1=O

Specifications

Boiling Point 111°C
Quantity 10 mL
Formula Weight 140.18
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetylcyclohexanone