missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbutyrolactone, 99+%

Catalog No. p-49474
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC102491000 100 mL
AC102492500 250 mL
AC102490010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC102491000 Supplier Thermo Scientific Chemicals Supplier No. 102491000
Only null left

CAS: 517-23-7 | C6H8O3 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 517-23-7
Molecular Formula C6H8O3
Molecular Weight (g/mol) 128.13
MDL Number MFCD00005394
InChI Key OMQHDIHZSDEIFH-UHFFFAOYNA-N
Synonym 2-acetylbutyrolactone, 3-acetyldihydrofuran-2 3h-one, alpha-acetylbutyrolactone, alpha-acetyl-gamma-butyrolactone, 3-acetyldihydro-2 3h-furanone, 2 3h-furanone, 3-acetyldihydro, 3-acetyltetrahydrofuran-2-one, 2-oxo-3-acetyltetrahydrofuran, 3-acetyltetrahydro-2-furanone, 3-acetyl-2 3h-4,5-dihydrofuranone
PubChem CID 10601
IUPAC Name 3-acetyloxolan-2-one
SMILES CC(=O)C1CCOC1=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Density 1.1800g/mL
Boiling Point 107°C to 108°C (5.0 mmHg)
Flash Point 140°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4585
Quantity 100 mL
Specific Gravity 1.18
Merck Index 15, 78
Solubility Information Solubility in water: 310g/L (20°C)
Formula Weight 128.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Acetylbutyrolactone, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-235-2
TSCA TSCA

RUO – Research Use Only