missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzoic Acid 98.0+%, TCI America™

Catalog No. A10755G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10755G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A10755G Supplier TCI America Supplier No. A10755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 577-56-0
Melting Point 118°C
Color White-Yellow
Molecular Formula C9H8O3
Quantity 5 g
MDL Number MFCD00002475
Synonym o-acetylbenzoic acid, benzoic acid, 2-acetyl, 2-acetylbenzoicacid, acetophenone-2-carboxylic acid, 2-acetyl-benzoic acid, benzoic acid, acetyl, 3-hydroxy-3-methylphthalide, 2'-acetophenonecarboxylic acid, acetophenone-2'-carboxylic acid, methyl phenyl ketone-o-carboxylic acid
InChI Key QDAWXRKTSATEOP-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC=C1C(O)=O
IUPAC Name 2-acetylbenzoic acid
Molecular Weight (g/mol) 164.16
PubChem CID 68474
Formula Weight 164.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetylbenzoic Acid
Show More Show Less