missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzo[b]furan, 99%

Catalog No. p-7046272
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1377106 5 g
AAL1377114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1377106 Supplier Thermo Scientific Chemicals Supplier No. L1377106

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1646-26-0 | C10H8O2 | 160.17 g/mol

2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylbenzo[b]furan is used as a fine chemical intermediate.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 1646-26-0
Molecular Formula C10H8O2
Molecular Weight (g/mol) 160.17
MDL Number MFCD00005849
InChI Key YUTFQTAITWWGFH-UHFFFAOYSA-N
Synonym 2-acetylbenzofuran, 1-benzofuran-2-yl ethanone, ethanone, 1-2-benzofuranyl, benzofuran-2-yl methyl ketone, 2-acetylcoumarone, 2-acetylcumarone, 2-benzofuranyl methyl ketone, benzo b furan-2-yl methyl ketone, 1-2-benzofuranyl ethanone, 2-acetylbenzo b furan
PubChem CID 15435
SMILES CC(=O)C1=CC2=CC=CC=C2O1

Specifications

Melting Point 70°C to 72°C
Boiling Point 110°C to 113°C (3 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 117910
Solubility Information Insoluble in water.
IUPAC Name 1-(1-benzofuran-2-yl)ethan-1-one
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 2-Acetylbenzo[b]furan
EINECSNumber 216-706-9
RTECSNumber OB1203000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only