missing translation for 'onlineSavingsMsg'
Learn More

2-Acetylbenzeneboronic acid, 97%

Catalog No. p-7029817
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2732706 5 g
AAH2732714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2732706 Supplier Thermo Scientific Chemicals Supplier No. H2732706
Only null left

CAS: 308103-40-4 | C8H9BO3 | 163.97 g/mol

2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetylphenylboronic acid is a reactant used for a variety of coupling reactions, as well as synthesis and hydroxy masking, Suzuki-Miyaura cross-coupling reactions, rhodium-catalyzed annulation of ynamides, transient masking of hydroxy groups, cationic palladium complex-catalyzed diastereoselective tandem annulation, hydrogen peroxide mediated formation of heteroaryl ethers and copper-catalyzed transmetalation and homocoupling.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 308103-40-4
Molecular Formula C8H9BO3
Molecular Weight (g/mol) 163.97
MDL Number MFCD01321263
InChI Key ZKAOVABYLXQUTI-UHFFFAOYSA-N
Synonym 2-acetylphenyl boronic acid, boronic acid, 2-acetylphenyl, 2'-boronoacetophenone, 1-2-boronophenyl ethan-1-one, 2-acetylbenzeneboronic acid, 2-acetylbenzenboronicacid, 1-2-dihydroxyboranyl phenyl ethan-1-one, 2-acetylphenylboronicacid, pubchem5373, acmc-1agh6
PubChem CID 2734309
IUPAC Name (2-acetylphenyl)boronic acid
SMILES CC(=O)C1=CC=CC=C1B(O)O

Specifications

Melting Point ∽170°C (decomposition)
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 163.97
Percent Purity 97%
Chemical Name or Material 2-Acetylbenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No

RUO – Research Use Only