missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-6-bromopyridine 98.0+%, TCI America™

Catalog No. A24041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24041G 1 g
A24045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24041G Supplier TCI America Supplier No. A24041G

Chemical Identifiers

CAS 49669-13-8
Molecular Formula C7H6BrNO
Molecular Weight (g/mol) 200.035
MDL Number MFCD00272200
InChI Key RUJTWTUYVOEEFW-UHFFFAOYSA-N
Synonym 2-acetyl-6-bromopyridine, 2-bromo-6-acetylpyridine, 1-6-bromopyridin-2-yl ethanone, 1-6-bromopyridin-2-yl ethan-1-one, 1-6-bromo-2-pyridinyl-1-ethanone, ethanone, 1-6-bromo-2-pyridinyl, bromopyridinylethanone, pubchem13351, acmc-1ap21, ksc490s1f
PubChem CID 11298578
IUPAC Name 1-(6-bromopyridin-2-yl)ethanone
SMILES CC(=O)C1=NC(=CC=C1)Br

Specifications

Melting Point 53°C
Quantity 1 g
Formula Weight 200.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-6-bromopyridine