missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-chlorothiophene 99.0+%, TCI America™

Catalog No. p-7117459
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
A14715G 5 g
A147125G 25 g
2 options

Catalog No. A14715G

Supplier: TCI America A14715G

Only null left

Chemical Identifiers

CAS 6310-09-4
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
PubChem CID 80572
IUPAC Name 1-(5-chlorothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Cl

Specifications

Melting Point 46°C
Boiling Point 118°C
Quantity 5 g
Formula Weight 160.62
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetyl-5-chlorothiophene
RTECSNumber OB1745000
TSCA No