missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-chlorothiophene, 99%

Catalog No. p-7081744
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1185009 10 g
AAA1185018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1185009 Supplier Thermo Scientific Chemicals Supplier No. A1185009
Only null left

CAS: 6310-09-4 | C6H5ClOS | 160.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6310-09-4
Molecular Formula C6H5ClOS
Molecular Weight (g/mol) 160.62
MDL Number MFCD00005444
InChI Key HTZGPEHWQCRXGZ-UHFFFAOYSA-N
Synonym 2-acetyl-5-chlorothiophene, 1-5-chlorothiophen-2-yl ethanone, 1-5-chloro-2-thienyl ethanone, 5-chloro-2-acetylthiophene, ethanone, 1-5-chloro-2-thienyl, 5-chlor-2-acetyl thiophen, ketone, 5-chloro-2-thienyl methyl, 1-5-chlorothiophen-2-yl ethan-1-one, 5-chloro-2-thienyl methyl ketone, 2-chloro-5-acetylthiophene
PubChem CID 80572
IUPAC Name 1-(5-chlorothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Cl

Specifications

Melting Point 44°C to 48°C
Boiling Point 117°C to 118°C (17 mmHg)
Flash Point 108°C (226°F)
Quantity 10 g
UN Number UN2811
Beilstein 113936
Sensitivity Light sensitive
Formula Weight 160.62
Percent Purity 99%
Chemical Name or Material 2-Acetyl-5-chlorothiophene
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H312-H332
Toxic if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
P261-P280-P301+P310-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 228-630-3
RTECSNumber OB1745000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only