missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-5-bromothiophene, 99%

Catalog No. AAA1482609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1482609 10 g
AAA1482618 50 g
AAA1482630 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1482609 Supplier Thermo Scientific Chemicals Supplier No. A1482609
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5370-25-2 | C6H5BrOS | 205.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5370-25-2
Molecular Formula C6H5BrOS
Molecular Weight (g/mol) 205.069
MDL Number MFCD00014528
InChI Key IGBZCOWXSCWSHO-UHFFFAOYSA-N
Synonym 2-acetyl-5-bromothiophene, 1-5-bromothiophen-2-yl ethanone, ethanone, 1-5-bromo-2-thienyl, 1-5-bromo-2-thienyl ethan-1-one, 1-5-bromothiophen-2-yl ethan-1-one, 5-bromo-2-thienyl methyl ketone, 2-acetyl-5-bromo thiophene, ketone, 5-bromo-2-thienyl methyl, 2-acetyl-5-bromo-thiophene, 1-5-bromo-2-thienyl ethanone
PubChem CID 79335
IUPAC Name 1-(5-bromothiophen-2-yl)ethanone
SMILES CC(=O)C1=CC=C(S1)Br

Specifications

Melting Point 93°C to 96°C
Boiling Point 103°C (4 mmHg)
Quantity 10 g
Beilstein 113949
Sensitivity Light sensitive
Formula Weight 205.07
Percent Purity 99%
Chemical Name or Material 2-Acetyl-5-bromothiophene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 226-363-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only