missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-methylpyrazine 98.0+%, TCI America™

Catalog No. A19781G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A19781G Supplier TCI America Supplier No. A19781G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23787-80-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00014612
InChI Key QUNOTZOHYZZWKQ-UHFFFAOYSA-N
Synonym 2-acetyl-3-methylpyrazine, 1-3-methylpyrazin-2-yl ethanone, 1-3-methylpyrazin-2-yl ethan-1-one, ethanone, 1-3-methylpyrazinyl, 2-methyl-3-acetylpyrazine, unii-7u5tew2y2b, 2-acetyl-3-methyl pyrazine, 1-3-methylpyrazinyl ethan-1-one, 3-acetyl-2-methylpyrazine, ethanone, 1-3-methyl-2-pyrazinyl
PubChem CID 32093
IUPAC Name 1-(3-methylpyrazin-2-yl)ethan-1-one
SMILES CC(=O)C1=NC=CN=C1C

Specifications

Boiling Point 69°C
Quantity 1 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetyl-3-methylpyrazine