missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3-aminothiophene, 97%

Catalog No. aal10563md
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL10563MD 250 mg
AAL1056303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL10563MD Supplier Thermo Scientific Chemicals Supplier No. L10563MD
Only null left

CAS: 31968-33-9 | C6H7NOS | 141.188 g/mol

Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dithienopyridines was synthesised from 2-Acetyl-3-aminothiophene. Also used as fine Intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Chemical Identifiers

CAS 31968-33-9
Molecular Formula C6H7NOS
Molecular Weight (g/mol) 141.188
MDL Number MFCD00052585
InChI Key MMLSKDGCLMDHAJ-UHFFFAOYSA-N
Synonym 2-acetyl-3-aminothiophene, 1-3-aminothiophen-2-yl ethanone, ethanone, 1-3-amino-2-thienyl, ethanone,1-3-amino-2-thienyl, pubchem20830, acmc-1co94, 1-3-aminothien-2-yl ethanone, 2-acetylthiophene-3-amine, 1-3-amino-2-thiophenyl ethanone
PubChem CID 2735233
IUPAC Name 1-(3-aminothiophen-2-yl)ethanone
SMILES CC(=O)C1=C(C=CS1)N

Specifications

Melting Point 84°C to 88°C
Boiling Point 120°C to 125°C (0.8 mmHg)
Quantity 250 mg
Beilstein 1635973
Solubility Information Insoluble in water.
Formula Weight 141.19
Percent Purity 97%
Chemical Name or Material 2-Acetyl-3-aminothiophene
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only