missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-methylpyrrole 98.0+%, TCI America™

Catalog No. A10145ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A10145ML 5 mL
A101425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A10145ML Supplier TCI America Supplier No. A10145ML

Chemical Identifiers

CAS 932-16-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00003089
InChI Key NZFLWVDXYUGFAV-UHFFFAOYSA-N
Synonym 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole
PubChem CID 61240
ChEBI CHEBI:59982
IUPAC Name 1-(1-methyl-1H-pyrrol-2-yl)ethan-1-one
SMILES CN1C=CC=C1C(C)=O

Specifications

Boiling Point 202°C
Quantity 5 mL
Formula Weight 123.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Acetyl-1-methylpyrrole