missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-1-methylpyrrole, 98%

Catalog No. AAA1535306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1535306 5 g
AAA1535314 25 g
AAA1535322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1535306 Supplier Thermo Scientific Chemicals Supplier No. A1535306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 932-16-1 | C7H9NO | 123.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 932-16-1
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.16
MDL Number MFCD00003089
InChI Key NZFLWVDXYUGFAV-UHFFFAOYSA-N
Synonym 2-acetyl-1-methylpyrrole, n-methyl-2-acetylpyrrole, 1-1-methyl-1h-pyrrol-2-yl ethanone, 1-1-methylpyrrol-2-yl ethanone, 1-methyl-2-acetylpyrrole, 2-acetyl-n-methylpyrrole, ethanone, 1-1-methyl-1h-pyrrol-2-yl, methyl 1-methylpyrrol-2-yl ketone, 1-1-methyl-1h-pyrrol-2-yl ethan-1-one, 2-acetyl-1-methyl pyrrole
PubChem CID 61240
ChEBI CHEBI:59982
IUPAC Name 1-(1-methylpyrrol-2-yl)ethanone
SMILES CN1C=CC=C1C(C)=O

Specifications

Density 1.04
Boiling Point 200°C to 202°C
Flash Point 68°C (154°F)
Odor Pleasant
Refractive Index 1.542
Quantity 5 g
Beilstein 111887
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 2-Acetyl-1-methylpyrrole
Hazard Category H227
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P280-P370+P378q-P501c
EINECSNumber 213-247-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only