missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetoxypropionyl Chloride 97.0+%, TCI America™

$294.81 - $882.52
Chemical Identifiers
CAS | 38939-83-2 |
---|---|
Molecular Formula | C5H7ClO3 |
Molecular Weight (g/mol) | 150.558 |
MDL Number | MFCD18969061 |
InChI Key | ALHZEIINTQJLOT-UHFFFAOYSA-N |
Synonym | O-Acetyllactoyl Chloride |
PubChem CID | 4547492 |
IUPAC Name | (1-chloro-1-oxopropan-2-yl) acetate |
SMILES | CC(C(=O)Cl)OC(=O)C |
Chemical Identifiers
38939-83-2 | |
150.558 | |
ALHZEIINTQJLOT-UHFFFAOYSA-N | |
4547492 | |
CC(C(=O)Cl)OC(=O)C |
C5H7ClO3 | |
MFCD18969061 | |
O-Acetyllactoyl Chloride | |
(1-chloro-1-oxopropan-2-yl) acetate |
Specifications
38939-83-2 | |
58°C | |
MFCD18969061 | |
3265 | |
ALHZEIINTQJLOT-UHFFFAOYSA-N | |
(1-chloro-1-oxopropan-2-yl) acetate | |
4547492 | |
≥97.0% (GC,T) | |
2-Acetoxypropionyl Chloride |
Yellow | |
C5H7ClO3 | |
1 g | |
O-Acetyllactoyl Chloride | |
CC(C(=O)Cl)OC(=O)C | |
150.558 | |
150.56 | |
Liquid |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator