missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxyisobutyryl Chloride 97.0+%, TCI America™

Catalog No. A11205G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A11205G 5 g
A112025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A11205G Supplier TCI America Supplier No. A11205G

Chemical Identifiers

CAS 40635-66-3
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.585
MDL Number MFCD00000708
InChI Key RBTCRFLJLUNCLL-UHFFFAOYSA-N
Synonym 2-acetoxyisobutyryl chloride, 1-chlorocarbonyl-1-methylethyl acetate, 2-acetoxy-2-methylpropionyl chloride, propanoyl chloride, 2-acetyloxy-2-methyl, 2-acetoxyisobutyroyl chloride, 2-chloro-1,1-dimethyl-2-oxoethyl acetate, acmc-1an8z, 2-acetoxyisobutyrylchloride, acetoxyisobutyric acid chloride, acetoxy-iso-butyrylchloride
PubChem CID 123496
IUPAC Name (1-chloro-2-methyl-1-oxopropan-2-yl) acetate
SMILES CC(=O)OC(C)(C)C(=O)Cl

Specifications

Color Yellow
Boiling Point 70°C
Quantity 5 g
UN Number 3265
Formula Weight 164.59
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-Acetoxyisobutyryl Chloride