missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxyisobutyryl bromide, 96%

Catalog No. AAL1159222
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1159222 100 g
AAL1159214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1159222 Supplier Thermo Scientific Chemicals Supplier No. L1159222
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 40635-67-4 | C6H9BrO3 | 209.039 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 40635-67-4
Molecular Formula C6H9BrO3
Molecular Weight (g/mol) 209.039
MDL Number MFCD00040923
InChI Key OOKAXSHFTDPZHP-UHFFFAOYSA-N
Synonym 2-acetoxyisobutyryl bromide, 2-acetoxy-2-methylpropionyl bromide, 1-bromocarbonyl-1-methylethyl acetate, alpha-acetoxy-isobutyryl bromide, a-acetoxyisobutyryl bromide, alpha-acetoxyisobutyryl bromide, 2-acetoxyisobutyryl bromide;2-acetoxy-2-methylpropionyl bromide, propanoyl bromide, 2-acetyloxy-2-methyl, aibb, pubchem17437
PubChem CID 2724715
IUPAC Name (1-bromo-2-methyl-1-oxopropan-2-yl) acetate
SMILES CC(=O)OC(C)(C)C(=O)Br

Specifications

Density 1.42
Boiling Point 100°C to 110°C (25 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.457
Quantity 100 g
UN Number UN3265
Beilstein 2432585
Sensitivity Moisture sensitive
Formula Weight 209.05
Percent Purity 96%
Chemical Name or Material 2-Acetoxyisobutyryl bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only