missing translation for 'onlineSavingsMsg'
Learn More

2-Acetoxycinnamic Acid 95.0+%, TCI America™

Catalog No. A24135G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24135G 5 g
A24131G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24135G Supplier TCI America Supplier No. A24135G

Chemical Identifiers

CAS 55620-18-3
Molecular Formula C11H10O4
Molecular Weight (g/mol) 206.197
MDL Number MFCD00014375
InChI Key UXOWQQCLBQBRMQ-VOTSOKGWSA-N
Synonym tylmarin, o-acetoxycinnamic acid, 2-acetoxycinnamic acid, o-acetyl-o-coumaric acid, cinnamic acid, 2-acetoxy, cinnamic acid, o-hydroxy-, acetate, 3-2-acetyloxy phenyl-2-propenoic acid, e-3-2-acetyloxyphenyl prop-2-enoic acid, 2-propenoic acid, 3-2-acetyloxy phenyl
PubChem CID 5372859
IUPAC Name (E)-3-(2-acetyloxyphenyl)prop-2-enoic acid
SMILES CC(=O)OC1=CC=CC=C1C=CC(=O)O

Specifications

Melting Point 156°C
Color White
Quantity 5 g
Formula Weight 206.20
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetoxycinnamic Acid
RTECSNumber GD7875000
TSCA No