missing translation for 'onlineSavingsMsg'
Learn More

2'-Acetoxyacetophenone 98.0+%, TCI America™

Catalog No. A19955G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19955G 5 g
A199525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19955G Supplier TCI America Supplier No. A19955G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7250-94-4
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00017223
InChI Key WWEDAIJJBDFOOK-UHFFFAOYSA-N
Synonym 2'-acetoxyacetophenone, 2-acetylphenyl acetate, ethanone, 1-2-acetyloxy phenyl, o-acetoxyacetophenone, 2'-acetoxy acetophenone, maybridge1_002653, acmc-209on8, acetic acid 2-acetyl-phenyl ester, acetophenone, 2-hydroxy-, acetate 8ci
PubChem CID 16716
IUPAC Name 2-acetylphenyl acetate
SMILES CC(=O)OC1=CC=CC=C1C(C)=O

Specifications

Melting Point 90°C
Boiling Point 118°C
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Acetoxyacetophenone