missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidothiazole 98.0+%, TCI America™

$160.30 - $561.08

Chemical Identifiers

CAS 2719-23-5
Molecular Formula C5H6N2OS
Molecular Weight (g/mol) 142.176
MDL Number MFCD00016892
InChI Key WXPLRSVMGRAIGW-UHFFFAOYSA-N
Synonym 2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide
PubChem CID 17625
IUPAC Name N-(1,3-thiazol-2-yl)acetamide
SMILES CC(=O)NC1=NC=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A14725G
View Documents
TCI America
A14725G
5 g
Each for $160.30
Only null left
 
A147225G
View Documents
TCI America
A147225G
25 g
Each for $561.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2719-23-5
142.176
WXPLRSVMGRAIGW-UHFFFAOYSA-N
17625
CC(=O)NC1=NC=CS1
C5H6N2OS
MFCD00016892
2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide
N-(1,3-thiazol-2-yl)acetamide

Specifications

2719-23-5
White-Yellow
MFCD00016892
2-acetamidothiazole, 2-acetylaminothiazole, n-thiazol-2-yl acetamide, acetamide, n-2-thiazolyl, n-1,3-thiazol-2-yl acetamide, n-2-thiazolylacetamide, thiazole, 2-acetamido, 2-acetaminothiazole, 2-acetamido-1,3-thiazole, n-thiazol-2-ylacetamide
CC(=O)NC1=NC=CS1
142.176
142.18
Crystalline Powder
207°C
C5H6N2OS
5 g
WXPLRSVMGRAIGW-UHFFFAOYSA-N
N-(1,3-thiazol-2-yl)acetamide
17625
≥98.0% (GC)
2-Acetamidothiazole
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0207

RTECSNumber : AC8800000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'