missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidoacrylic Acid 98.0+%, TCI America™

Catalog No. A14435G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A14435G Supplier TCI America Supplier No. A14435G

Chemical Identifiers

CAS 5429-56-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00004257
InChI Key UFDFFEMHDKXMBG-UHFFFAOYSA-N
Synonym 2-acetamidoacrylic acid, 2-propenoic acid, 2-acetylamino, acrylic acid, 2-acetamido, 2-acetamidoacrylicacid, acetyldehydroalanine, n-acetyldehydroalanine, 2-acetylamino acrylic acid, .alpha.-acetamidoacrylic acid, 2-acetylamino-2-propenoic acid, 2-acetylamino prop-2-enoic acid
PubChem CID 79482
IUPAC Name 2-acetamidoprop-2-enoic acid
SMILES CC(=O)NC(=C)C(=O)O

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 129.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamidoacrylic Acid
TSCA No
Recommended Storage Refrigerator