missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamidoacrylic acid, 97+%

Catalog No. AAL0164906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0164906 5 g
AAL0164914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0164906 Supplier Thermo Scientific Chemicals Supplier No. L0164906
Only null left

CAS: 5429-56-1 | C5H7NO3 | 129.115 g/mol

2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Acetamidoacrylic acid is used as an antioxidant as well as a monomer.

Solubility
Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 5429-56-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.115
MDL Number MFCD00004257
InChI Key UFDFFEMHDKXMBG-UHFFFAOYSA-N
Synonym 2-acetamidoacrylic acid, 2-propenoic acid, 2-acetylamino, acrylic acid, 2-acetamido, 2-acetamidoacrylicacid, acetyldehydroalanine, n-acetyldehydroalanine, 2-acetylamino acrylic acid, .alpha.-acetamidoacrylic acid, 2-acetylamino-2-propenoic acid, 2-acetylamino prop-2-enoic acid
PubChem CID 79482
IUPAC Name 2-acetamidoprop-2-enoic acid
SMILES CC(=O)NC(=C)C(=O)O

Specifications

Melting Point ∽190°C (decomposition)
Quantity 5 g
Beilstein 1812032
Solubility Information Soluble in dimethyl sulfoxide. Slightly soluble in water and methanol.
Formula Weight 129.12
Percent Purity 99%
Chemical Name or Material 2-Acetamidoacrylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-583-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only