missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Acetamido-6-hydroxypurine 95.0+%, TCI America™
$336.90 - $1345.69
Chemical Identifiers
| CAS | 19962-37-9 |
|---|---|
| Molecular Formula | C7H7N5O2 |
| Molecular Weight (g/mol) | 193.17 |
| MDL Number | MFCD00078201 |
| InChI Key | MXSMRDDXWJSGMC-UHFFFAOYSA-N |
| Synonym | 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide |
| PubChem CID | 88319 |
| IUPAC Name | N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide |
| SMILES | CC(=O)NC1=NC(=O)C2=C(N1)N=CN2 |
Chemical Identifiers
| 19962-37-9 | |
| 193.17 | |
| MXSMRDDXWJSGMC-UHFFFAOYSA-N | |
| 88319 | |
| CC(=O)NC1=NC(=O)C2=C(N1)N=CN2 |
| C7H7N5O2 | |
| MFCD00078201 | |
| 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide | |
| N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide |
Specifications
| 19962-37-9 | |
| C7H7N5O2 | |
| 1 g | |
| MXSMRDDXWJSGMC-UHFFFAOYSA-N | |
| N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide | |
| 88319 | |
| ≥95.0% (HPLC,T) | |
| 2-Acetamido-6-hydroxypurine |
| White-Yellow | |
| MFCD00078201 | |
| 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide | |
| CC(=O)NC1=NC(=O)C2=C(N1)N=CN2 | |
| 193.17 | |
| 193.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No
Recommended Storage : Freezer