missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-6-hydroxypurine 95.0+%, TCI America™

Catalog No. A12481G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12481G 1 g
A12485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A12481G Supplier TCI America Supplier No. A12481G

Chemical Identifiers

CAS 19962-37-9
Molecular Formula C7H7N5O2
Molecular Weight (g/mol) 193.17
MDL Number MFCD00078201
InChI Key MXSMRDDXWJSGMC-UHFFFAOYSA-N
Synonym 2-acetamido-6-hydroxypurine, n-2-acetylguanine, n2-acetylguanine, n-acetylguanine, n-6-hydroxy-9h-purin-2-yl acetamide, 2-n-acetylguanine, acetamide, n-6,7-dihydro-6-oxo-1h-purin-2-yl, n-6,7-dihydro-6-oxo-1h-purin-2-yl acetamide, n-6-hydroxypurin-2-yl acetamide, n-6-oxo-1,9-dihydropurin-2-yl acetamide
PubChem CID 88319
IUPAC Name N-(6-oxo-6,7-dihydro-3H-purin-2-yl)acetamide
SMILES CC(=O)NC1=NC(=O)C2=C(N1)N=CN2

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 193.17
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-6-hydroxypurine
TSCA No
Recommended Storage Freezer