missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl Azide 98.0+%, TCI America™

Catalog No. A16161G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16161G 1 g
A16165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A16161G Supplier TCI America Supplier No. A16161G

Chemical Identifiers

CAS 6205-69-2
Molecular Formula C14H20N4O8
Molecular Weight (g/mol) 372.33
MDL Number MFCD00216968
InChI Key RMCFMPMNMQZHSF-UHFFFAOYNA-N
PubChem CID 2725015
IUPAC Name [3,4-bis(acetyloxy)-6-azido-5-acetamidooxan-2-yl]methyl acetate
SMILES CC(=O)NC1C(OC(COC(C)=O)C(OC(C)=O)C1OC(C)=O)N=[N+]=[N-]

Specifications

Melting Point 167°C
Quantity 1 g
Formula Weight 372.33
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-3,4,6-tri-O-acetyl-2-deoxy-beta-D-glucopyranosyl Azide
TSCA No
Recommended Storage Freezer