missing translation for 'onlineSavingsMsg'
Learn More

2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-beta-D-glucopyranose 98.0+%, TCI America™

Catalog No. p-7117451
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A14591G 1 g
A14595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A14591G Supplier TCI America Supplier No. A14591G
Only null left

Chemical Identifiers

CAS 7772-79-4
Molecular Formula C16H23NO10
Molecular Weight (g/mol) 389.357
MDL Number MFCD00006595
InChI Key OVPIZHVSWNOZMN-OXGONZEZSA-N
Synonym n-acetyl-beta-d-glucosamine tetraacetate, 2-acetamido-1,3,4,6-tetra-o-acetyl-2-deoxy-beta-d-glucopyranose, beta-d-glucosamine pentaacetate, 2-acetamido-2-deoxy-beta-d-glucopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-5-acetamido-3,4,6-triacetoxy-tetrahydropyran-2-yl methyl acetate, beta-d-galactosamine pentaacetate, 2-acetylamino-2-deoxy-beta-d-galactopyranose 1,3,4,6-tetraacetate, 2r,3s,4r,5r,6s-3,4,6-tris acetyloxy-5-acetamidooxan-2-yl methyl acetate, n-acetyl-, a-d-glucosamine tetraacetate, glc-n-ac free
PubChem CID 99461
IUPAC Name [(2R,3S,4R,5R,6S)-5-acetamido-3,4,6-triacetyloxyoxan-2-yl]methyl acetate
SMILES CC(=O)NC1C(C(C(OC1OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 1 g
Formula Weight 389.36
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-1,3,4,6-tetra-O-acetyl-2-deoxy-beta-D-glucopyranose
TSCA No
Recommended Storage Freezer