missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-5-decyne 97.0+%, TCI America™

Catalog No. D11935ML
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11935ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11935ML Supplier TCI America Supplier No. D11935ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 19550-56-2
Molecular Formula C12H22
Molecular Weight (g/mol) 166.308
MDL Number MFCD00041611
InChI Key RCFYJIGZQAOTFV-UHFFFAOYSA-N
Synonym 2,9-dimethyl-5-decyne, 2,9-dimethyl-dec-5-yne, acmc-1bu20, 5-decyne, 2,9-dimethyl, rcfyjigzqaotfv-uhfffaoysa
PubChem CID 140552
IUPAC Name 2,9-dimethyldec-5-yne
SMILES CC(C)CCC#CCCC(C)C

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 mL
Formula Weight 166.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,9-Dimethyl-5-decyne