missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-5-decyne, 96%, Thermo Scientific Chemicals

Catalog No. p-7035690
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5341403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5341403 Supplier Thermo Scientific Chemicals Supplier No. H5341403

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19550-56-2 | Molecular Formula: C12H22 | Molecular Weight: 166.308 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19550-56-2
Molecular Formula C12H22
Molecular Weight (g/mol) 166.308
MDL Number MFCD00041611
InChI Key RCFYJIGZQAOTFV-UHFFFAOYSA-N
Synonym 2,9-dimethyl-5-decyne, 2,9-dimethyl-dec-5-yne, acmc-1bu20, 5-decyne, 2,9-dimethyl, rcfyjigzqaotfv-uhfffaoysa
PubChem CID 140552
IUPAC Name 2,9-dimethyldec-5-yne
SMILES CC(C)CCC#CCCC(C)C

Specifications

Density 0.77
Boiling Point 92°C (19 mmHg)
Linear Formula (CH3)2CH(CH2)2C≡C(CH2)2CH(CH3)2
Quantity 1 g
Formula Weight 166.31
Percent Purity 96%
Chemical Name or Material 2,9-Dimethyl-5-decyne
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only