missing translation for 'onlineSavingsMsg'
Learn More

2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline, 98%

Catalog No. AC161340010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161340010 1 g Glass bottle
AC161340050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161340010 Supplier Thermo Scientific Chemicals Supplier No. 161340010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4733-39-5 | C26H20N2 | 360.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4733-39-5
Molecular Formula C26H20N2
Molecular Weight (g/mol) 360.46
MDL Number MFCD00004972
InChI Key STTGYIUESPWXOW-UHFFFAOYSA-N
Synonym bathocuproine, bathocuproin, 1,10-phenanthroline, 2,9-dimethyl-4,7-diphenyl, unii-9thp2v94fx, 9thp2v94fx, 2,9-dimethyl-4,7-diphenylpyridino 3,2-h quinoline, pubchem19657, bathocuproin 1g, bidd:gt0563
PubChem CID 65149
IUPAC Name 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline
SMILES CC1=CC(C2=CC=CC=C2)=C2C=CC3=C(C=C(C)N=C3C2=N1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 277°C to 285°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Beilstein 23, IV, 2160
Merck Index 15, 1005
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,2-propanol,chloroform,benzene
Formula Weight 360.45
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline
Show More Show Less
EINECSNumber 225-240-5

RUO – Research Use Only