missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,9-Diacetylguanine, 96%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 459210250
Specifications
| 2,9-Diacetylguanine | |
| 285.0°C | |
| C9H9N5O3 | |
| 25 g | |
| GILZZWCROUGLIS-UHFFFAOYSA-N | |
| N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide | |
| 76461 | |
| 96% |
| 3056-33-5 | |
| Authentic | |
| MFCD00142116 | |
| n2,9-diacetylguanine, n,9-diacetylguanine, n 2 ,9-diacetylguanine, 2,9-diacetylguanine, 2,9-diacetateguanine, n-9-acetyl-6-oxo-6,9-dihydro-1h-purin-2-yl acetamide, unii-vuw2b9yr95, 2,9-diacetyl guanine, vuw2b9yr95, n-9-acetyl-6-oxo-3h-purin-2-yl acetamide | |
| CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O | |
| 235.20 | |
| 235.2 |
Chemical Identifiers
| 3056-33-5 | |
| 235.20 | |
| GILZZWCROUGLIS-UHFFFAOYSA-N | |
| N-(9-acetyl-6-oxo-3H-purin-2-yl)acetamide |
| C9H9N5O3 | |
| MFCD00142116 | |
| 76461 | |
| CC(=O)NC1=NC(=O)C2=C(N1)N(C=N2)C(C)=O |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 221-287-
RUO – Research Use Only