missing translation for 'onlineSavingsMsg'
Learn More

2-(9-Carbazolyl)ethylboronic acid pinacol ester, 98%

Catalog No. AAL19580MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL19580MD 250 mg
AAL1958003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL19580MD Supplier Thermo Scientific Chemicals Supplier No. L19580MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 608534-41-4 | C20H24BNO2 | 321.227 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 608534-41-4
Molecular Formula C20H24BNO2
Molecular Weight (g/mol) 321.227
MDL Number MFCD03788738
InChI Key NTUZDLWSQJRXAM-UHFFFAOYSA-N
Synonym 2-9-carbazolyl ethylboronic acid pinacol ester, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl-9h-carbazole, 9-2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 9-2-tetramethyl-1,3,2-dioxaborolan-2-yl ethyl carbazole, 2-2-9h-carbazole-9-yl ethyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 45925651
IUPAC Name 9-[2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)CCN2C3=CC=CC=C3C4=CC=CC=C42

Specifications

Melting Point 111°C to 113°C
Quantity 250 mg
Formula Weight 321.23
Percent Purity 98%
Chemical Name or Material 2-(9-Carbazolyl)ethylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only