missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dimethylanthra[2,3-b:6,7-b']dithiophene (purified by sublimation), TCI America™

Catalog No. d4617100mg
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4617100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4617100MG Supplier TCI America Supplier No. D4617100MG
Only null left

Chemical Identifiers

CAS 1019983-99-3
Molecular Formula C20H14S2
Molecular Weight (g/mol) 318.452
InChI Key HPJACFAPBXKLQA-UHFFFAOYSA-N
Synonym anti-DMADT
PubChem CID 88958401
SMILES CC1=CC2=CC3=CC4=CC5=C(C=C(S5)C)C=C4C=C3C=C2S1

Specifications

Quantity 100 mg
Formula Weight 318.45
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dimethylanthra[2,3-b:6,7-b′]dithiophene (purified by sublimation)