missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,8-Diiododibenzofuran 98.0+%, TCI America™
$156.59 - $533.13
Chemical Identifiers
| CAS | 5943-11-3 |
|---|---|
| Molecular Formula | C12H6I2O |
| Molecular Weight (g/mol) | 419.99 |
| MDL Number | MFCD00094363 |
| InChI Key | VKTLGHDNPWVFSY-UHFFFAOYSA-N |
| PubChem CID | 634525 |
| IUPAC Name | 2,8-diiododibenzofuran |
| SMILES | C1=CC2=C(C=C1I)C3=C(O2)C=CC(=C3)I |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D4835200MG
|
TCI America
D4835200MG |
200 mg |
Each for $156.59
|
|
|||||
|
D48351G
|
TCI America
D48351G |
1 g |
Each for $533.13
|
|
|||||
Chemical Identifiers
| 5943-11-3 | |
| 419.99 | |
| VKTLGHDNPWVFSY-UHFFFAOYSA-N | |
| 2,8-diiododibenzofuran |
| C12H6I2O | |
| MFCD00094363 | |
| 634525 | |
| C1=CC2=C(C=C1I)C3=C(O2)C=CC(=C3)I |
Specifications
| 5943-11-3 | |
| C12H6I2O | |
| 200 mg | |
| C1=CC2=C(C=C1I)C3=C(O2)C=CC(=C3)I | |
| 419.99 | |
| 419.99 | |
| Crystalline Powder |
| 176°C | |
| MFCD00094363 | |
| VKTLGHDNPWVFSY-UHFFFAOYSA-N | |
| 2,8-diiododibenzofuran | |
| 634525 | |
| ≥98.0% (GC) | |
| 2,8-Diiododibenzofuran |
Safety and Handling
TSCA : No