missing translation for 'onlineSavingsMsg'
Learn More

2,8-Dibromodibenzofuran 98.0+%, TCI America™

Catalog No. p-7128485
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48211G 1 g
D48215G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48211G Supplier TCI America Supplier No. D48211G
Only null left

Chemical Identifiers

CAS 10016-52-1
Molecular Formula C12H6Br2O
Molecular Weight (g/mol) 325.99
MDL Number MFCD00093683
InChI Key UFCZRCPQBWIXTR-UHFFFAOYSA-N
PubChem CID 82290
IUPAC Name 4,12-dibromo-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2(7),3,5,10,12-hexaene
SMILES BrC1=CC2=C(OC3=C2C=C(Br)C=C3)C=C1

Specifications

Melting Point 195°C
Color White-Yellow
Quantity 1 g
Formula Weight 325.99
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Dibromodibenzofuran