missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 2-8 CELLsium™ Medium for Short-Term Cell Storage

2-8 CELLsius is a cytoprotective, protein free, ready to use bio-solution without DMSO for the short-term storage of cells or tissue.

$503.10

Specifications

Form Liquid
Sterility Sterile
For Use With (Application) Short-term cell storage
Content And Storage Stored at 2 to 8°C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MP092780349
View Documents Promotion Details
MP Biomedicals™
092780349
100 mL
Each for $503.10
Only null left
 
Description

Description

  • For short-term storage, add cold (2 to 8 °C) 2-8 CELLsium at a recommended cell concentration of 1 x 106 mL to 8 x 106 mL
  • The 2-8 CELLsium can be removed after centrifugation, cell concentration adjusted and transferred to an appropriate culture flask with growth medium
  • It will keep the cells healthy while in a temporary senescent phase, unlike regular growth media, which cannot
Specifications

Specifications

Liquid
Sterile
Short-term cell storage
Stored at 2 to 8°C
Videos
SDS
Documents

Documents