missing translation for 'onlineSavingsMsg'
Learn More

2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%

CAS: 50816-20-1 | C13H25BrO2 | 293.245 g/mol

$101.67 - $340.70

Chemical Identifiers

CAS 50816-20-1
Molecular Formula C13H25BrO2
Molecular Weight (g/mol) 293.245
MDL Number MFCD00014648
InChI Key JCRBYQZIJFWGOO-UHFFFAOYSA-N
Synonym 2-8-bromooctyl oxy tetrahydro-2h-pyran, 2h-pyran, 2-8-bromooctyl oxy tetrahydro, 1-bromo-8-tetrahydropyranyloxy octane, 8-bromooctyl tetrahydropyranyl ether, 1-bromo-8-tetrahydropyranyloxyoctane, 2-8-bromooctyloxy tetrahydropyran, 2-8-bromooctyl oxy oxane, acmc-20aowo
PubChem CID 170916
IUPAC Name 2-(8-bromooctoxy)oxane
SMILES C1CCOC(C1)OCCCCCCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0108106
View Documents
Thermo Scientific Chemicals
L0108106
5 g
Each for $101.67
Only null left
 
AAL0108114
View Documents
Thermo Scientific Chemicals
L0108114
25 g
Each for $340.70
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50816-20-1
293.245
JCRBYQZIJFWGOO-UHFFFAOYSA-N
170916
C1CCOC(C1)OCCCCCCCCBr
C13H25BrO2
MFCD00014648
2-8-bromooctyl oxy tetrahydro-2h-pyran, 2h-pyran, 2-8-bromooctyl oxy tetrahydro, 1-bromo-8-tetrahydropyranyloxy octane, 8-bromooctyl tetrahydropyranyl ether, 1-bromo-8-tetrahydropyranyloxyoctane, 2-8-bromooctyloxy tetrahydropyran, 2-8-bromooctyl oxy oxane, acmc-20aowo
2-(8-bromooctoxy)oxane

Specifications

50816-20-1
>110°C (230°F)
1.477
5 g
2-8-bromooctyl oxy tetrahydro-2h-pyran, 2h-pyran, 2-8-bromooctyl oxy tetrahydro, 1-bromo-8-tetrahydropyranyloxy octane, 8-bromooctyl tetrahydropyranyl ether, 1-bromo-8-tetrahydropyranyloxyoctane, 2-8-bromooctyloxy tetrahydropyran, 2-8-bromooctyl oxy oxane, acmc-20aowo
C1CCOC(C1)OCCCCCCCCBr
293.245
293.25
Technical
130°C to 132°C (0.8 mmHg)
C13H25BrO2
MFCD00014648
1306644
JCRBYQZIJFWGOO-UHFFFAOYSA-N
2-(8-bromooctoxy)oxane
170916
90%
2-(8-Bromooctyloxy)tetrahydropyran
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 256-786-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only