missing translation for 'onlineSavingsMsg'
Learn More

2-(8-Bromooctyloxy)tetrahydropyran, tech. 90%

Catalog No. AAL0108106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0108106 5 g
AAL0108114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0108106 Supplier Thermo Scientific Chemicals Supplier No. L0108106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 50816-20-1 | C13H25BrO2 | 293.245 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 50816-20-1
Molecular Formula C13H25BrO2
Molecular Weight (g/mol) 293.245
MDL Number MFCD00014648
InChI Key JCRBYQZIJFWGOO-UHFFFAOYSA-N
Synonym 2-8-bromooctyl oxy tetrahydro-2h-pyran, 2h-pyran, 2-8-bromooctyl oxy tetrahydro, 1-bromo-8-tetrahydropyranyloxy octane, 8-bromooctyl tetrahydropyranyl ether, 1-bromo-8-tetrahydropyranyloxyoctane, 2-8-bromooctyloxy tetrahydropyran, 2-8-bromooctyl oxy oxane, acmc-20aowo
PubChem CID 170916
IUPAC Name 2-(8-bromooctoxy)oxane
SMILES C1CCOC(C1)OCCCCCCCCBr

Specifications

Boiling Point 130°C to 132°C (0.8 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.477
Quantity 5 g
Beilstein 1306644
Formula Weight 293.25
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-(8-Bromooctyloxy)tetrahydropyran
EINECSNumber 256-786-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only