missing translation for 'onlineSavingsMsg'
Learn More

2,8-Bis(trifluoromethyl)-4-hydroxyquinoline 98.0+%, TCI America™

Catalog No. B37141G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B37141G Supplier TCI America Supplier No. B37141G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 35853-41-9
Melting Point 133°C
Molecular Formula C11H5F6NO
Quantity 1 g
MDL Number MFCD00075091
Synonym 2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
InChI Key JIWHKBAFGFPZKM-UHFFFAOYSA-N
SMILES C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F
IUPAC Name 2,8-bis(trifluoromethyl)-1H-quinolin-4-one
Molecular Weight (g/mol) 281.157
PubChem CID 736140
Formula Weight 281.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline
Show More Show Less