missing translation for 'onlineSavingsMsg'
Learn More

2,8-Bis(trifluoromethyl)-4-hydroxyquinoline, 97%, Thermo Scientific Chemicals

Catalog No. p-7024669
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1975006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1975006 Supplier Thermo Scientific Chemicals Supplier No. A1975006

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 35853-41-9 | Molecular Formula: C11H5F6NO | Molecular Weight: 281.157 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 35853-41-9
Molecular Formula C11H5F6NO
Molecular Weight (g/mol) 281.157
MDL Number MFCD00075091
InChI Key JIWHKBAFGFPZKM-UHFFFAOYSA-N
Synonym 2,8-bis trifluoromethyl-4-hydroxyquinoline, 2,8-bis trifluoromethyl quinolin-4-ol, 2,8-bis trifluoromethyl-4-quinolinol, 2,8-bis trifluoromethyl-1h-quinolin-4-one, 2,8-bis-trifluoromethyl-quinolin-4-ol, cc-pmlsc-dma-p104, 4-quinolinol, 2,8-bis trifluoromethyl, 2,8-bis trifluoromethyl-4-hydroxy quinoleine, 2,8-di trifluoromethyl quinolin-4-ol, pubchem5934
PubChem CID 736140
IUPAC Name 2,8-bis(trifluoromethyl)-1H-quinolin-4-one
SMILES C1=CC2=C(C(=C1)C(F)(F)F)NC(=CC2=O)C(F)(F)F

Specifications

Melting Point 129°C to 135°C
Quantity 5 g
Formula Weight 281.16
Percent Purity 97%
Chemical Name or Material 2,8-Bis(trifluoromethyl)-4-hydroxyquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-762-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only