missing translation for 'onlineSavingsMsg'
Learn More

2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. B54895G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B54895G 5 g
B54891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B54895G Supplier TCI America Supplier No. B54895G
Only null left

Chemical Identifiers

CAS 1270030-08-4
Molecular Formula C26H29BO2
Synonym 2-(tert-Butyl)-7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrene, [7-(tert-Butyl)pyren-2-yl]boronic Acid Pinacol Ester

Specifications

Melting Point 241°C
Color White-Yellow
Quantity 5 g
Formula Weight 384.33
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-[7-(tert-Butyl)pyren-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane