missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Octadienol (cis- and trans- mixture) 95.0+%, TCI America™
$72.70 - $212.51
Chemical Identifiers
| CAS | 23578-51-0 |
|---|---|
| Molecular Formula | C8H14O |
| Molecular Weight (g/mol) | 126.199 |
| MDL Number | MFCD00191472 |
| InChI Key | YHYGSIBXYYKYFB-VOTSOKGWSA-N |
| PubChem CID | 5365650 |
| IUPAC Name | (2E)-octa-2,7-dien-1-ol |
| SMILES | C=CCCCC=CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O024325ML
|
TCI America
O024325ML |
25 mL |
Each for $72.70
|
|
|||||
|
O0243500ML
|
TCI America
O0243500ML |
500 mL |
Each for $212.51
|
|
|||||
Chemical Identifiers
| 23578-51-0 | |
| 126.199 | |
| YHYGSIBXYYKYFB-VOTSOKGWSA-N | |
| (2E)-octa-2,7-dien-1-ol |
| C8H14O | |
| MFCD00191472 | |
| 5365650 | |
| C=CCCCC=CCO |
Specifications
| 23578-51-0 | |
| C8H14O | |
| 25 mL | |
| C=CCCCC=CCO | |
| 126.199 | |
| 126.20 | |
| Liquid |
| Yellow | |
| MFCD00191472 | |
| YHYGSIBXYYKYFB-VOTSOKGWSA-N | |
| (2E)-octa-2,7-dien-1-ol | |
| 5365650 | |
| ≥95.0% (GC) | |
| 2,7-Octadienol (cis- and trans- mixture) |
Safety and Handling
RTECSNumber : RG5425000
TSCA : No