missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,7-Octadienol (cis- and trans- mixture) 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Formula Weight | 126.20 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,7-Octadienol (cis- and trans- mixture) |
Safety and Handling
| RTECSNumber | RG5425000 |
| TSCA | No |