missing translation for 'onlineSavingsMsg'
Learn More

2,7-Octadienol (cis- and trans- mixture) 95.0+%, TCI America™

Catalog No. O024325ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O024325ML 25 mL
O0243500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O024325ML Supplier TCI America Supplier No. O024325ML
Only null left

Chemical Identifiers

CAS 23578-51-0
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00191472
InChI Key YHYGSIBXYYKYFB-VOTSOKGWSA-N
PubChem CID 5365650
IUPAC Name (2E)-octa-2,7-dien-1-ol
SMILES C=CCCCC=CCO

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 126.20
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,7-Octadienol (cis- and trans- mixture)
RTECSNumber RG5425000
TSCA No