missing translation for 'onlineSavingsMsg'
Learn More

2,7-Diphenyl-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7128766
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54271G 1 g
D54275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54271G Supplier TCI America Supplier No. D54271G
Only null left

Chemical Identifiers

CAS 42448-04-4
Molecular Formula C24H17N
Molecular Weight (g/mol) 319.407
InChI Key NWLCIOKUOGGKKK-UHFFFAOYSA-N
PubChem CID 66717301
IUPAC Name 2,7-diphenyl-9H-carbazole
SMILES C1=CC=C(C=C1)C2=CC3=C(C=C2)C4=C(N3)C=C(C=C4)C5=CC=CC=C5

Specifications

Melting Point 345°C
Color White-Yellow
Quantity 1 g
Formula Weight 319.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diphenyl-9H-carbazole